Läs mer
Bepotastine >90%, TRC
Produktkod. 30339520
Change view
Kvantitet:
5 mg
Produktkod. Kvantitet
30339520 5 mg
1 options
Produktkod. 30339520 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB3160955MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Bepotastine
CAS 125602-71-3
Molekylformel C21 H25 Cl N2 O3
Kvantitet 5 mg
Synonym (+)-(S)-4-[4-[1-(4-chlorophenyl)-1-(2-pyridyl)methoxy]piperidin-1-yl]butyric acid, 4-[4-[(S)-(4-chlorophenyl)-pyridin-2-ylmethoxy]piperidin-1-yl]butanoic acid, 1-Piperidinebutanoic acid, 4-[(S)-(4-chlorophenyl)-2-pyridinylmethoxy], 4-[(S)-(4-Chlorophenyl)-2-pyridinylmethoxy]-1-piperidinebutanoic acid, (S)-4-[4-(4-chlorophenyl-2-pyridylmethoxy)piperidinyl]butyric acid, (S)-Bepotastine, Bepotastine
InChI formel InChI=1S/C21H25ClN2O3/c22-17-8-6-16(7-9-17)21(19-4-1-2-12-23-19)27-18-10-14-24(15-11-18)13-3-5-20(25)26/h1-2,4,6-9,12,18,21H,3,5,10-11,13-15H2,(H,25,26)/t21-/m0/s1
LEDER OC(=O)CCCN1CCC(CC1)O[C@@H](c2ccc(Cl)cc2)c3ccccn3
IUPAC-namn 4-[4-[(S)-(4-chlorophenyl)-(2-pyridyl)methoxy]-1-piperidyl]butanoic acid
Molekylvikt (g/mol) 388.89
Formel vikt 388.1554