Läs mer
Bepotastine Ethyl Ester, TRC
Produktkod. 30341935
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30341935 100 mg
1 options
Produktkod. 30341935 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB317005100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material Bepotastine Ethyl Ester
CAS 190730-39-3
Anteckningar om renhetsgrad HPLC
Molekylformel C23H29ClN2O3
Kvantitet 100 mg
Synonym (S)-4-[4-(4-Chlorophenyl-2-pyridylmethoxy)piperidinyl]butyric Acid Ethyl Ester, (S)-4-[(4-Chlorophenyl)-2-pyridinylmethoxy]-1-piperidinebutanoic Acid Ethyl Ester, 4-[(S)-(4-Chlorophenyl)-2-pyridinylmethoxy]-1-piperidinebutanoic Acid Ethyl Ester
InChI formel InChI=1S/C23H29ClN2O3/c1-2-28-22(27)7-5-15-26-16-12-20(13-17-26)29-23(21-6-3-4-14-25-21)18-8-10-19(24)11-9-18/h3-4,6,8-11,14,20,23H,2,5,7,12-13,15-17H2,1H3/t23-/m0/s1
LEDER ClC1=CC=C([C@H](OC2CCN(CCCC(OCC)=O)CC2)C3=CC=CC=N3)C=C1
IUPAC-namn ethyl 4-[4-[(S)-(4-chlorophenyl)-pyridin-2-ylmethoxy]piperidin-1-yl]butanoate
Molekylvikt (g/mol) 416.94
Formel vikt 416.1867