Läs mer
Berberine Chloride Hydrate, TRC
Produktkod. 30137305
Change view
Kvantitet:
10 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137305 25 g
30135515 100 g
17998594 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137305 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB31815025G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 633-65-8
Molekylformel C20 H18 N O4 . Cl
Molekylvikt (g/mol) 371.81
Synonym Benzo[g]-1,3-benzodioxolo[5,6-a]quinolizinium, 5,6-dihydro-9,10-dimethoxy-, chloride, Berberine, chloride, Berbinium, 7,8,13,13a-tetradehydro-9,10-dimethoxy-2,3-(methylenedioxy)-, chloride, Berberine hydrochloride, Berberinium chloride, NSC 163088, NSC 646666
IUPAC-namn 5,6-Dihydro-9,10-dimethoxybenzo[g]-1,3-benzodioxolo[5,6-a]quinolizinium chloride
LEDER [Cl-].COc1ccc2cc3c4cc5OCOc5cc4CC[n+]3cc2c1OC
Specifikationer
Kemiskt namn eller material Berberine chloride
Anteckningar om renhetsgrad HPLC
Kvantitet 25 g
InChI formel InChI=1S/C20H18NO4.ClH/c1-22-17-4-3-12-7-16-14-9-19-18(24-11-25-19)8-13(14)5-6-21(16)10-15(12)20(17)23-2;/h3-4,7-10H,5-6,11H2,1-2H3;1H/q+1;/p-1
Formel vikt 371.0924
Procent renhet >95%