missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bestrophin-3 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20443-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
BEST3 belongs to the bestrophin family of anion channels, which includes BEST1, the gene mutant in vitelliform macular dystrophy (VMD), and 2 other BEST1-like genes, BEST2 and BEST4. Bestrophins are transmembrane (TM) proteins that share a homology region containing a high content of aromatic residues, including an invariant arg-phe-pro (RFP) motif. The bestrophin genes share a conserved gene structure, with almost identical sizes of the 8 RFP-TM domain-encoding exons and highly conserved exon-intron boundaries. Each of the 4 bestrophin genes has a unique 3-prime end of variable length.Specifications
Bestrophin-3 | |
Polyclonal | |
Unconjugated | |
BEST3 | |
BEST3, Bestrophin 3, Bestrophin-3, VMD2L3 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
144453, 382427 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6H1V1, Q8N1M1 | |
Best3 | |
Bestrophin-3 Fusion Protein Ag14255 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |