missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ beta-2 Microglobulin Monoclonal Antibody (246-E8.E7 (HLA-ABC-m2))
GREENER_CHOICE

Produktkod. 11354843
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
500 μL
Förpackningsstorlek:
500µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11354843 500 μL 500µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11354843 Leverantör Invitrogen™ Leverantörsnummer MA513418

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Mouse Monoclonal Antibody

MA5-13418 targets Microglobulin Beta-2 in FACS and IF applications and shows reactivity with Human samples. The MA5-13418 immunogen is human PBL (T-ALL patient).

Beta-2-microglobulin (B2M) is a 12-14 kDa protein that is a subunit of the major histocompatibility complex (MHC) class I. Human beta 2 Microglobulin associated with HLA Class I antigens are expressed on many types of cells including lymphocytes, thymocytes, monocytes, granulocytes, platelets, endothelial cells, and epithelial cells, but is absent on erythrocytes. Beta-2 microglobulin is also found in free form in the plasma. The amino acid sequence of Beta-2-microglobulin is homologous to the constant regions of immunoglobulin heavy and light chains, with a predominantly beta pleated sheet structure. The protein is ubiquitously expressed in the cytoplasm, and is involved in antigen processing and presentation. Beta-2-microglobulin also plays a role in TCR signaling, and downstream signaling in naive CD8+ cells. Beta 2-microglobulin is encoded by the B2M gene located on chromosome 15. A mutation in the Beta-2-microglobulin gene has been shown to result in hypercatabolic hypoproteinemia. Levels of beta 2 microglobulin can be used to assess renal function in kidney transplant recipients. Beta-2-microglobulin that protein can be used as a prognostic marker for HIV infection.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen beta-2 Microglobulin
Användningsområden Flow Cytometry, Immunocytochemistry
Klassificering Monoclonal
Klona 246-E8.E7 (HLA-ABC-m2)
Koncentration 0.2 mg/mL
Konjugera Unconjugated
Formulering PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gen B2m
Genaccessionsnr. P61769
Gene Alias B2m; B2-microglobulin; b2m-W01; b2m-W03; b2m-Z01; b2m-Z02; b2m-Z03; Beta 2 microglobin; Beta 2 microglobulin; beta 2-microglobulin; beta chain of MHC class I molecules; Beta2 microglobulin; beta-2 microglobulin; beta-2 microglobulin preprotein; beta2m; beta2-m; beta-2-microglobin; beta-2-microglobulin; beta-2-microglobulin C-terminal fragment (55 AA); Beta-2-microglobulin form pI 5.3; B-F; CDABP0092; HDCMA22P; IMD43; Lactollin; Ly-m11; MHC Class I; microglobulin beta-2; Orla-B2m
Gensymboler B2m
Värd art Mouse
Immunogen Human PBL (T-ALL patient).
Reningsmetod Protein A
Kvantitet 500 μL
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 567
Målarter Human
Innehåll och lagring 4°C
Produkttyp Antibody
Form Liquid
Isotyp IgG2a κ
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.