Läs mer
beta-(4-Chlorophenoxy)-alpha-(1,1-dimethylethyl)-1H-1,2,4-triazole-1-ethanol(Mixture of Diastereomers) (Triadimenol), TRC
Produktkod. 30362447
Change view
Kvantitet:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30362447 10 g
30364631 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30362447 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC37506010G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 55219-65-3
Molekylformel C14 H18 Cl N3 O2
Molekylvikt (g/mol) 295.76
Synonym Bayfidan, Bayfidan EW, Baytan, Baytan 15, Baytan N, Baytan TF 3479B, KWG 0519, Photon 60GR, Shavit, Spinnaker, Triadimenol, UK 199, Vydine, β-(4-Chlorophenoxy)-α-(1,1-dimethylethyl)-1H-1,2,4-triazole-1-ethanol
IUPAC-namn 1-(4-chlorophenoxy)-3,3-dimethyl-1-(1,2,4-triazol-1-yl)butan-2-ol
LEDER CC(C)(C)C(O)C(Oc1ccc(Cl)cc1)n2cncn2
Specifikationer
Kemiskt namn eller material Triadimenol
Kvantitet 10 g
InChI formel InChI=1S/C14H18ClN3O2/c1-14(2,3)12(19)13(18-9-16-8-17-18)20-11-6-4-10(15)5-7-11/h4-9,12-13,19H,1-3H3
Formel vikt 295.1088