Läs mer
Thermo Scientific™ beta-Casomorphin (1-3), Amide
Produktkod. 15417199
Kvantitet:
5 mg
25 mg
Förpackningsstorlek:
25mg
5mg
Produktkod. 15417199
Brand: Thermo Scientific™ J66008.#0
Request Bulk or Custom Format
Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter
For cellular and molecular biology applications
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifikationer
Reningsmetod HPLC
Innehåll och lagring -30°C to -10°C
Formatera Solid
Molekylvikt (g/mol) 424.49
Produkttyp beta-Casomorphin (1-3), amide
Kvantitet 5 mg
Regulatorisk status RUO - research use only
Sekvens Tyr-Pro-Phe-NH2
For Research Use Only