Läs mer
Invitrogen™ beta Catenin Monoclonal Antibody (15B8), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
16850.00 SEK - 38090.00 SEK
Specifikationer
| Antigen | beta Catenin |
|---|---|
| Klona | 15B8 |
| Koncentration | 0.5 mg/mL |
| Användningsområden | Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
| Klassificering | Monoclonal |
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Kvantitet | Pris | Kvantitet och tillgänglighet | |||||
|
17170612
|
Invitrogen™
14-2567-37 |
2 mg |
16850.00 SEK
2mg |
Vänligen Logga in för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag! | |||||
|
17180612
|
Affymetrix eBioscience
14-2567-95 |
10 mg | |||||||
Beskrivning
The 15B8 MAb reacts with human and mouse beta-catenin, one member of a family of catenins, which are intracellular proteins that interact with cadherins to mediate cellular adhesion. More specifically, beta-catenin binds to the cytoplasmic tail of E-cadherin. In addition, this molecule is a component of the canonical Wnt signaling pathway. In the absence of Wnt binding its receptor, beta-catenin is phosphorylated and resides in the cytoplasm where it is eventually targeted for degradation by ubiquitination. Upon Wnt binding, beta-catenin becomes dephosphorylated, translocates to the nucleus, and modulates gene expression in partnership with the transcription factors T cell factor (TCF) and lymphocyte enhancer binding factor (LEF). Expression of beta-catenin is found in a wide variety of non-immune and immune tissues, including thymocytes and T and B lymphocytes. The Wnt and beta-catenin signaling pathway has been demonstrated to play a crucial role in the development of T, B, and hematopoietic stem cells.
Beta-catenin, an adherens junction (AJ) protein, was originally identified as a component of cell-cell adhesion structures. AJs are necessary for the creation and maintenance of epithelial cell layers by regulating cell growth and adhesion between cells. Beta-catenin interacts with the cytoplasmic domain of E-cadherin and links E-cadherin to alpha-catenin, which in turn mediates anchorage of the E-cadherin complex to the cortical actin cytoskeleton. Studies show that Beta-catenin also binds to another cytoskeletal complex containing the adenomatous polyposis coli protein and microtubules, and interacts with several signaling pathways that include tyrosine kinases, phosphatases and Wnt/Wingless. The interplay between beta-catenin, cytoskeletal complexes and signaling pathways may regulate morphogenesis. Beta-catenin is expressed in several hair follicle cell types, basal and peripheral matrix cells, and cells of the outer and inner root sheats. A pathological role of beta-catenin has been identified in pilomatrixoma (PTR), medulloblastoma (MDB), colorectal cancer (CRC), ovarian cancer, and tumor development. In the nucleus, beta-catenin serves to co activate a family of Lef/Tcf transcription factors that stimulate transcription of target genes including those encoding cyclin D and c-myc that promote cell proliferation. The influence on cell proliferation is the molecular basis for the role of beta-catenin in tumorgenesis, specifically, solid tumors of the breast, colon, liver, lung, gastric, prostate, and skin.Specifikationer
| beta Catenin | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| 12387, 1499 | |
| Primary | |
| 4°C | |
| CTNNB1 |
| 15B8 | |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Mouse | |
| Human, Mouse | |
| P35222, Q02248 | |
| IgG1 κ | |
| Affinity chromatography | |
| Antibody |
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.