Läs mer
Beta-Chlorolactic Acid, TRC
Produktkod. 17940675
Change view
Kvantitet:
500 mg
1 g
2.5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17940675 2.5 g
17970995 1 g
17930885 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17940675 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC3676702.5G
Denna vara kräver en prisuppdatering innan den beställs.
Begär ett pris för denna vara här.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1713-85-5
Molekylformel C3 H5 Cl O3
Molekylvikt (g/mol) 124.523
Synonym 3-Chloro-lactic Acid, (±)-3-Chloro-2-hydroxypropanoic Acid, (±)-3-Chlorolactic Acid, 2-Hydroxy-3-chloropropionic Acid, 3-Chloro-2-hydroxypropanoic Acid, 3-Chloro-2-hydroxypropionic Acid, 3-Chlorolactic Acid, Chlorolactic Acid, DL-3-Chlorolactic Acid, DL-β-Chlorolactic Acid, 3-Chloro-2-hydroxy-propionic Acid, 3-Chloro-2-hydroxy-propanoic Acid
IUPAC-namn 3-chloro-2-hydroxy-propanoic acid
LEDER OC(CCl)C(=O)O
Specifikationer
Kemiskt namn eller material Beta-Chlorolactic Acid
Kvantitet 2.5 g
InChI formel InChI=1S/C3H5ClO3/c4-1-2(5)3(6)7/h2,5H,1H2,(H,6,7)
Formel vikt 123.9927