missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
Beta-Cortol, MedChemExpress
Beskrivning
Beta-Cortol is an androgen metabolite present in adults.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Beta-Cortol |
| CAS | 667-65-2 |
| Färg | White |
| Anteckningar om renhetsgrad | Research |
| Molekylformel | C21H36O5 |
| Kvantitet | 5 mg |
| Löslighetsinformation | DMSO : 100 mg/mL (271.36 mM; Need ultrasonic) |
| LEDER | C[C@]1(C[C@@H]2O)[C@](CC[C@]1(O)[C@H](O)CO)([H])[C@@]3([H])[C@@]2([H])[C@](CC[C@@H](O)C4)(C)[C@]4([H])CC3 |
| Molekylvikt (g/mol) | 368.51 |
| Formel vikt | 368.51 |
| Visa mer |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?