Läs mer
Beta-Cyanobenzenepropanoic Acid Ethyl Ester, TRC
Produktkod. 30348895
Change view
Kvantitet:
250 mg
Produktkod. Kvantitet
30348895 250 mg
1 options
Produktkod. 30348895 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC962355250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material beta-Cyanobenzenepropanoic Acid Ethyl Ester
CAS 14025-83-3
Molekylformel C12 H13 N O2
Kvantitet 250 mg
Synonym Ethyl 3-(3-Cyanophenyl)propanoate, Ethyl 3-cyano-3-phenylpropanoate, Ethyl 3-cyano-3-phenylpropionate, Ethyl beta-cyanohydrocinnamate
InChI formel InChI=1S/C12H13NO2/c1-2-15-12(14)8-11(9-13)10-6-4-3-5-7-10/h3-7,11H,2,8H2,1H3
LEDER CCOC(=O)CC(C#N)c1ccccc1
IUPAC-namn ethyl 3-cyano-3-phenylpropanoate
Molekylvikt (g/mol) 203.237
Formel vikt 203.095