Läs mer
Beta-Cyanobenzenepropanoic Acid Ethyl Ester, TRC
Produktkod. 30366826
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30366826 1 g
30360021 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30366826 Leverantör Toronto Research Chemicals Leverantörsnummer TRCC9623551G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material beta-Cyanobenzenepropanoic Acid Ethyl Ester
CAS 14025-83-3
Molekylformel C12 H13 N O2
Kvantitet 1 g
Synonym Ethyl 3-(3-Cyanophenyl)propanoate, Ethyl 3-cyano-3-phenylpropanoate, Ethyl 3-cyano-3-phenylpropionate, Ethyl beta-cyanohydrocinnamate
InChI formel InChI=1S/C12H13NO2/c1-2-15-12(14)8-11(9-13)10-6-4-3-5-7-10/h3-7,11H,2,8H2,1H3
LEDER CCOC(=O)CC(C#N)c1ccccc1
IUPAC-namn ethyl 3-cyano-3-phenylpropanoate
Molekylvikt (g/mol) 203.237
Formel vikt 203.095