Läs mer
beta-Estradiol 17-Acetate, TRC
Produktkod. 30134159
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30134159 250 mg
30012566 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30134159 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE888005250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1743-60-8
Molekylformel C20 H26 O3
Molekylvikt (g/mol) 314.42
Synonym Estra-1,3,5(10)-triene-3,17-diol (17β)-, 17-acetate, Estradiol, 17-acetate (6CI,7CI,8CI), 17β-Acetoxyestra-1,3,5(10)-trien-3-ol, 17β-Acetylestradiol, 17β-Estradiol 17-acetate, Estra-1,3,5(10)-triene-3,17β-diol 17-acetate, Estradiol 17-monoacetate, β-Estradiol 17-acetate
IUPAC-namn [(8R,9S,13S,14S,17S)-3-hydroxy-13-methyl-6,7,8,9,11,12,14,15,16,17-decahydrocyclopenta[a]phenanthren-17-yl] acetate
LEDER CC(=O)O[C@H]1CC[C@H]2[C@@H]3CCc4cc(O)ccc4[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material 17-beta-Estradiol 17-acetate
Kvantitet 250 mg
InChI formel InChI=1S/C20H26O3/c1-12(21)23-19-8-7-18-17-5-3-13-11-14(22)4-6-15(13)16(17)9-10-20(18,19)2/h4,6,11,16-19,22H,3,5,7-10H2,1-2H3/t16-,17-,18+,19+,20+/m1/s1
Formel vikt 314.1882