Läs mer
Beta-Estradiol 17-(Beta-D-Glucuronide) Sodium Salt, TRC
Produktkod. 30004348
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004348 1 mg
30010802 5 mg
30005659 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30004348 Leverantör Toronto Research Chemicals Leverantörsnummer TRCE8889481MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 15087-02-2
Molekylformel C24H31NaO8
Molekylvikt (g/mol) 470.49
LEDER C[C@@]12[C@](CC[C@@H]2O[C@@H]3O[C@@H]([C@H]([C@@H]([C@H]3O)O)O)C([O])=O)([H])[C@@]4([H])[C@@](CC1)([H])C5=CC=C(O)C=C5CC4.[Na]
Specifikationer
Kemiskt namn eller material beta-Estradiol 17-(beta-D-Glucuronide) Sodium Salt
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
Procent renhet >95%