Läs mer
beta-Formylpropionic Acid, TRC
Produktkod. 30012554
Change view
Kvantitet:
2.5 mg
5 mg
Förpackningsstorlek:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30012554 2.5 mg 2.5mg
30005589 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30012554 Leverantör Toronto Research Chemicals Leverantörsnummer TRCF7009352.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 692-29-5
Molekylformel C4 H6 O3
Molekylvikt (g/mol) 102.09
Synonym Butanoic acid, 4-oxo-, Succinaldehydic acid (6CI,7CI,8CI), 4-Oxobutanoic acid, 3-Formylpropanoic acid, 3-Formylpropionic acid, 4-Oxobutyric acid, Butyraldehydic acid, Propanoic acid, 3-formyl-, Succinic acid semialdehyde, Succinic semialdehyde, β-Formylpropionic acid, γ-Oxybutyric acid
IUPAC-namn 4-oxobutanoic acid
LEDER OC(=O)CCC=O
Specifikationer
Kemiskt namn eller material beta-Formylpropionic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 mg
InChI formel InChI=1S/C4H6O3/c5-3-1-2-4(6)7/h3H,1-2H2,(H,6,7)
Formel vikt 102.0317
Procent renhet >95%