missing translation for 'onlineSavingsMsg'
Läs mer
Läs mer
beta Galactosidase Monoclonal Antibody (BGAL01), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA511935
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
MA5-11935 targets Beta-Galactosidase in immunoprecipitation and Western blot applications and shows reactivity with All samples. The MA5-11935 immunogen is e. coli beta-galactosidase.
Beta Galactosidase encodes a member of the glycosyl hydrolase 35 family of proteins. Alternative splicing results in multiple transcript variants, at least one of which encodes a preproprotein that is proteolytically processed to generate the mature lysosomal enzyme. This enzyme catalyzes the hydrolysis of a terminal beta-linked galactose residue from ganglioside substrates and other glycoconjugates. Mutations in this gene have been associated with GM1-gangliosidosis and Morquio B syndrome. Diseases associated with GLB1 include Gm1-Gangliosidosis, Type I and Gm1-Gangliosidosis, Type Ii.
Specifikationer
| beta Galactosidase | |
| Monoclonal | |
| 0.2 mg/mL | |
| PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Bacteria | |
| Antibody | |
| IgG1 |
| Immunoprecipitation, Western Blot | |
| BGAL01 | |
| Unconjugated | |
| acid beta-galactosidase; AW125515; betagal; Beta-Gal; Beta-galactosidase; bgal; Bge; Bgl; Bgl-e; Bgl-s; Bgl-t; Bgs; Bgt; C130097A14Rik; EBP; Elastin receptor 1; elastin receptor 1, 67kDa; ELNR1; galactosidase beta 1; galactosidase, beta 1; GLB1; Glb-1; lactase; lysosmal hydrolase; lysosomal beta-galactosidase; lysosomal hydrolase; MPS4B; βgal; β-gal; β-galactosidase | |
| E. coli beta-galactosidase | |
| 500 μL | |
| Primary | |
| 4° C | |
| Liquid |
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering