Läs mer
(+/-)-Beta-Hydrastine, TRC
Produktkod. 30407450
Change view
Kvantitet:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30407450 2.5 mg
30403051 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30407450 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH6756902.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 60594-55-0
Molekylformel C21H21NO6
Molekylvikt (g/mol) 383.39
Synonym (3S)-rel-6,7-Dimethoxy-3-[(5R)-5,6,7,8-tetrahydro-6-methyl-1,3-dioxolo[4,5-g]isoquinolin-5-yl]-1(3H)-isobenzofuranone, (+/-)-Hydrastine
LEDER CN1CCC2=CC3=C(C=C2[C@@H]1[C@@H]4C5=C(C(=C(C=C5)OC)OC)C(=O)O4)OCO3
Specifikationer
Kemiskt namn eller material (+/-)-beta-Hydrastine
Kvantitet 2.5 mg
InChI formel InChI=1S/C21H21NO6/c1-22-7-6-11-8-15-16(27-10-26-15)9-13(11)18(22)19-12-4-5-14(24-2)20(25-3)17(12)21(23)28-19/h4-5,8-9,18-19H,6-7,10H2,1-3H3/t18-,19+/m1/s1