Läs mer
Beta-Hydroxyl Phentermine, TRC
Produktkod. 30136210
Change view
Kvantitet:
10 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136210 25 mg
30005857 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30136210 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH95404025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 34405-42-0
Molekylformel C10H15NO
Molekylvikt (g/mol) 165.23
Synonym α-(1-Amino-1-methylethyl)-benzyl Alcohol, α-(1-Amino-1-methylethyl)benzenemethanol, (2-Hydroxy-1,1-dimethyl-2-phenylethyl)amine, (±)-α-(1-Methyl-1-aminoethyl)benzyl Alcohol, 2-Phenyl-1,1-dimethylethanolamine, NSC 17705;
IUPAC-namn 2-amino-2-methyl-1-phenylpropan-1-ol
LEDER OC(C(C)(N)C)C1=CC=CC=C1
Specifikationer
Kemiskt namn eller material beta-Hydroxyl Phentermine
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C10H15NO/c1-10(2,11)9(12)8-6-4-3-5-7-8/h3-7,9,12H,11H2,1-2H3
Formel vikt 165.1154
Procent renhet >95%