Läs mer
Beta-Hyodeoxycholic Acid, TRC
Produktkod. 30135575
Change view
Kvantitet:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30135575 50 mg
30006760 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30135575 Leverantör Toronto Research Chemicals Leverantörsnummer TRCH99810550MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 570-84-3
Molekylformel C24H40O4
Molekylvikt (g/mol) 392.57
Synonym (3beta,5beta,6alpha)-3,6-Dihydroxycholan-24-oic Acid, 3beta,6alpha-dihydroxy-5beta-Cholan-24-oic Acid, Isohyodeoxycholic Acid
IUPAC-namn (4R)-4-[(3S,5R,6S,8S,9S,10R,13R,14S,17R)-3,6-dihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
LEDER C[C@H](CCC(=O)O)[C@H]1CC[C@@H]2[C@@]1(CC[C@H]3[C@H]2C[C@@H]([C@H]4[C@@]3(CC[C@@H](C4)O)C)O)C
Specifikationer
Kemiskt namn eller material beta-Hyodeoxycholic Acid
Anteckningar om renhetsgrad HPLC
Kvantitet 50 mg
InChI formel InChI=1S/C24H40O4/c1-14(4-7-22(27)28)17-5-6-18-16-13-21(26)20-12-15(25)8-10-24(20,3)19(16)9-11-23(17,18)2/h14-21,25-26H,4-13H2,1-3H3,(H,27,28)/t14-,15+,16+,17-,18+,19+,20+,21+,23-,24-/m1/s1
Formel vikt 392.2927
Procent renhet >95%