Läs mer
Beta-Lumi (-)-Colchicine, TRC
Produktkod. 30005237
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30005237 10 mg
30005513 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30005237 Leverantör Toronto Research Chemicals Leverantörsnummer TRCL47430010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 6901-13-9
Molekylformel C22 H25 N O6
Molekylvikt (g/mol) 399.44
Synonym Colchicine Imp. C (EP), N-[(7S,7bR,10aS)-1,2,3,9-Tetramethoxy-8-oxo-5,6,7,7b,8,10a-hexahydrobenzo-[a]cyclopenta[3,4]cyclobuta[1,2-c]cyclohepten-7-yl]acetamide, N-[(7S,7bR,10aS)-5,6,7,7b,8,10a-Hexahydro-1,2,3,9-tetramethoxy-8-oxobenzo[a]cyclopenta[3,4]cyclobuta[1,2-c]cyclohepten-7-yl]acetamide, [7S-(7.alpha.,7bβ,10aβ)]-N-(5,6,7,7b,8,10a-Hexahydro-1,2,3,9-tetramethoxy-8-oxobenzo[a]cyclopenta[3,4]cyclobuta[1,2-c]cyclohepten-7-yl)acetamide, β-Lumicolchicine, Benzo[a]cyclopenta[3,4]cyclobuta[1,2-c]cycloheptene, acetamide deriv., (-)-β-Lumicolchicine, Lumicolchicine, NSC 221661, NSC 56233, β-Lumi-(-)-colchicine
IUPAC-namn N-[(7S,7bR,10aS)-1,2,3,9-Tetramethoxy-8-oxo-5,6,7,7b,8,10a-hexahydrobenzo-[a]cyclopenta[3,4]cyclobuta[1,2-c]cyclohepten-7-yl]acetamide
LEDER COC1=C[C@H]2[C@@H](C1=O)C3=C2c4c(CC[C@@H]3NC(=O)C)cc(OC)c(OC)c4OC
Specifikationer
Kemiskt namn eller material beta-Lumicolchicine
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C22H25NO6/c1-10(24)23-13-7-6-11-8-15(27-3)21(28-4)22(29-5)16(11)17-12-9-14(26-2)20(25)18(12)19(13)17/h8-9,12-13,18H,6-7H2,1-5H3,(H,23,24)/t12-,13+,18-/m1/s1
Formel vikt 399.1682
Procent renhet >95%