Läs mer
Beta-Methyl Digoxin, TRC
Produktkod. 30006539
Change view
Kvantitet:
1 mg
2.5 mg
10 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30006539 10 mg -
30008345 1 mg -
30010620 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30006539 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM30185010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 30685-43-9
Molekylformel C42H66O14
Molekylvikt (g/mol) 794.97
Synonym Card-20(22)-enolide, 3-[(O-2,6-dideoxy-4-O-methyl-β-D-ribo-hexopyranosyl-(1→4)-O-2,6-dideoxy-β-D-ribo-hexopyranosyl-(1→4)-2,6-dideoxy-β-D-ribo-hexopyranosyl)oxy]-12,14-dihydroxy-, (3β,5β,12β)-, Digoxin, 4' ' '-O-methyl- (8CI), (3β,5β,12β)-3-[(O-2,6-Dideoxy-4-O-methyl-β-D-ribo-hexopyranosyl-(1→4)-O-2,6-dideoxy-β-D-ribo-hexopyranosyl-(1→4)-2,6-dideoxy-β-D-ribo-hexopyranosyl)oxy]-12,14-dihydroxycard-20(22)-enolide, 4' ' '-Methyldigoxin, 4' ' '-O-Methyldigoxin, 4' ' '-β-Methyldigoxin, Lanirapid, Lanitop, Medigoxin, Methyldigoxin, Metildigoxin, β-Methyldigoxin
LEDER CO[C@H]1[C@@H](O)C[C@H](O[C@H]2[C@@H](O)C[C@H](O[C@H]3[C@@H](O)C[C@H](O[C@H]4CC[C@@]5(C)[C@H](CC[C@@H]6[C@@H]5C[C@@H](O)[C@]7(C)[C@H](CC[C@]67O)C8=CC(=O)OC8)C4)O[C@@H]3C)O[C@@H]2C)O[C@@H]1C
Specifikationer
Kemiskt namn eller material beta-Methyl Digoxin
Kvantitet 10 mg
InChI formel InChI=1S/C42H66O14/c1-20-37(49-6)29(43)16-35(51-20)55-39-22(3)53-36(18-31(39)45)56-38-21(2)52-34(17-30(38)44)54-25-9-11-40(4)24(14-25)7-8-27-28(40)15-32(46)41(5)26(10-12-42(27,41)48)23-13-33(47)50-19-23/h13,20-22,24-32,34-39,43-46,48H,7-12,14-19H2,1-6H3/t20-,21-,22-,24-,25+,26-,27-,28+,29+,30+,31+,32-,34+,35+,36+,37-,38-,39-,40+,41+,42+/m1/s1
Formel vikt 794.4453