Läs mer
Beta-Methylcholine Chloride, TRC
Produktkod. 30137028
Change view
Kvantitet:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30137028 5 g
30133909 10 g
30010126 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137028 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM3058555G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2382-43-6
Molekylformel C6H16ClNO
Molekylvikt (g/mol) 153.65
Synonym beta-Hydroxypropyltrimethylammonium Chloride, (2-Hydroxypropyl)trimethylammonium Chloride, 2-Hydroxy-N,N,N-trimethyl-1-propanaminium Chloride;
IUPAC-namn 2-hydroxypropyl(trimethyl)azanium;chloride
LEDER [Cl-].CC(O)C[N+](C)(C)C
Specifikationer
Kemiskt namn eller material beta-Methylcholine Chloride
Kvantitet 5 g
InChI formel InChI=1S/C6H16NO.ClH/c1-6(8)5-7(2,3)4;/h6,8H,5H2,1-4H3;1H/q+1;/p-1
Formel vikt 153.092