Läs mer
Beta-Muricholic Acid, TRC
Produktkod. 30011072
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011072 10 mg
30008081 1 mg
30012205 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011072 Leverantör Toronto Research Chemicals Leverantörsnummer TRCM73275510MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2393-59-1
Molekylformel C24 H40 O5
Molekylvikt (g/mol) 408.57
Synonym Cholan-24-oic acid, 3,6,7-trihydroxy-, (3α,5β,6β,7β)-, 5β-Cholan-24-oic acid, 3α,6β,7β-trihydroxy- (8CI), 5β-Cholanic acid, 3α,6β,7β-trihydroxy- (7CI), β-Muricholic acid (6CI), (3α,5β,6β,7β)-3,6,7-Trihydroxycholan-24-oic acid, 3α,6β,7β-Trihydroxy-5β-cholan-24-oic acid, 5β-Cholanic acid-3α,6β,7β-triol, 3α,6β,7β-Trihydroxy-5β-cholanic acid
IUPAC-namn (4R)-4-[(3R,5R,6S,7R,8S,9S,10R,13R,14S,17R)-3,6,7-trihydroxy-10,13-dimethyl-2,3,4,5,6,7,8,9,11,12,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl]pentanoic acid
LEDER C[C@H](CCC(=O)O)[C@H]1CC[C@H]2[C@@H]3[C@@H](O)[C@@H](O)[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
Specifikationer
Kemiskt namn eller material beta-Muricholic acid
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C24H40O5/c1-13(4-7-19(26)27)15-5-6-16-20-17(9-11-23(15,16)2)24(3)10-8-14(25)12-18(24)21(28)22(20)29/h13-18,20-22,25,28-29H,4-12H2,1-3H3,(H,26,27)/t13-,14-,15-,16+,17+,18+,20+,21+,22-,23-,24-/m1/s1
Formel vikt 408.2876
Procent renhet >95%