Läs mer
Beta-N-Benzyl-N'-methylaminopropiophenone, TRC
Produktkod. 30346466
Change view
Kvantitet:
1 g
Produktkod. Kvantitet
30346466 1 g
1 options
Produktkod. 30346466 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB6955551G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material beta-N-Benzyl-N'-methylaminopropiophenone
CAS 21970-65-0
Molekylformel C17H19NO
Kvantitet 1 g
Synonym 3-[Methyl(phenylmethyl)amino]-1-phenyl-1-propanone
InChI formel InChI=1S/C17H19NO/c1-18(14-15-8-4-2-5-9-15)13-12-17(19)16-10-6-3-7-11-16/h2-11H,12-14H2,1H3
LEDER CN(CCC(=O)c1ccccc1)Cc2ccccc2
IUPAC-namn 3-[benzyl(methyl)amino]-1-phenylpropan-1-one
Molekylvikt (g/mol) 253.34
Formel vikt 253.1467