missing translation for 'onlineSavingsMsg'
Läs mer

Beta-Naphthyl Beta-D-Glucopyranoside Tetraacetate, TRC

Produktkod. 30474925
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30474925 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30474925 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN368510250MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Kemiskt namn eller material beta-Naphthyl beta-D-Glucopyranoside Tetraacetate
CAS 14581-89-6
Molekylformel C24 H26 O10
Kvantitet 250 mg
InChI formel InChI=1S/C24H26O10/c1-13(25)29-12-20-21(30-14(2)26)22(31-15(3)27)23(32-16(4)28)24(34-20)33-19-10-9-17-7-5-6-8-18(17)11-19/h5-11,20-24H,12H2,1-4H3/t20-,21-,22+,23-,24-/m1/s1
LEDER CC(=O)OC[C@H]1O[C@@H](Oc2ccc3ccccc3c2)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C
IUPAC-namn [(2R,3R,4S,5R,6S)-3,4,5-triacetyloxy-6-naphthalen-2-yloxyoxan-2-yl]methyl acetate
Molekylvikt (g/mol) 474.46
Formel vikt 474.1526
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.