Läs mer
beta-Naphthyl beta-D-Glucopyranoside, TRC
Produktkod. 30483571
Change view
Kvantitet:
500 mg
Produktkod. Kvantitet
30483571 500 mg
1 options
Produktkod. 30483571 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN368505500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material 2-Naphthyl-beta-D-Glucopyranoside
CAS 6044-30-0
Molekylformel C16 H18 O6
Kvantitet 500 mg
Synonym 2-Naphthol glucoside, β-Naphthol glucoside
InChI formel InChI=1S/C16H18O6/c17-8-12-13(18)14(19)15(20)16(22-12)21-11-6-5-9-3-1-2-4-10(9)7-11/h1-7,12-20H,8H2/t12-,13-,14+,15-,16-/m1/s1
LEDER OC[C@H]1O[C@@H](Oc2ccc3ccccc3c2)[C@H](O)[C@@H](O)[C@@H]1O
IUPAC-namn (2R,3S,4S,5R,6S)-2-(hydroxymethyl)-6-(2-naphthyloxy)tetrahydropyran-3,4,5-triol
Molekylvikt (g/mol) 306.31
Formel vikt 306.1103