Läs mer
beta-Nicotyrine, TRC
Produktkod. 17980775
Change view
Kvantitet:
10 mg
25 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17980775 100 mg
17900675 25 mg
17992785 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17980775 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN445000100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 487-19-4
Molekylformel C10 H10 N2
Molekylvikt (g/mol) 158.2
Synonym Pyridine, 3-(1-methyl-1H-pyrrol-2-yl)-, Nicotyrine (6CI,7CI), Pyridine, 3-(1-methylpyrrol-2-yl)- (8CI), 1-Methyl-2-(3-pyridyl)pyrrole, 3,2'-Nicotyrine, 3-(1-Methyl-2-pyrrolyl)pyridine, NSC 127943, NSC 407276, β-Nicotyrine, beta-Nicotyrine
IUPAC-namn 3-(1-methylpyrrol-2-yl)pyridine
LEDER Cn1cccc1c2cccnc2
Specifikationer
Kemiskt namn eller material beta-Nicotyrine
Anteckningar om renhetsgrad HPLC
Kvantitet 100 mg
InChI formel InChI=1S/C10H10N2/c1-12-7-3-5-10(12)9-4-2-6-11-8-9/h2-8H,1H3
Formel vikt 158.0844
Procent renhet >95%