Läs mer
beta-Nornicotyrine, TRC
Produktkod. 30132679
Change view
Kvantitet:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30132679 10 mg
17982795 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30132679 Leverantör Toronto Research Chemicals Leverantörsnummer TRCN82500010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 494-98-4
Molekylformel C9H8N2
Molekylvikt (g/mol) 144.1732
Synonym Pyridine, 3-(1H-pyrrol-2-yl)-, Pyridine, 3-pyrrol-2-yl- (6CI,7CI,8CI), Nornicotyrine, β-Nornicotyrine
IUPAC-namn 3-(1H-pyrrol-2-yl)pyridine
LEDER c1cncc(c1)c2ccc[nH]2
Specifikationer
Kemiskt namn eller material 3-(1H-Pyrrol-2-yl)pyridine (beta-Nornicotyrine)
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C9H8N2/c1-3-8(7-10-5-1)9-4-2-6-11-9/h1-7,11H
Formel vikt 144.0687
Procent renhet >95%