Läs mer
Beta-Phellandrene, TRC
Produktkod. 17982455
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
17982455 1 g
17974505 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 17982455 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP2945601G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 555-10-2
Molekylformel C10 H16
Molekylvikt (g/mol) 136.23
Synonym Cyclohexene, 3-methylene-6-(1-methylethyl)-, p-Mentha-1(7),2-diene (8CI), 3-Methylene-6-(1-methylethyl)cyclohexene, (±)-β-Phellandrene, 3-Isopropyl-6-methylene-1-cyclohexene, 4-Isopropyl-1-methylene-2-cyclohexene, NSC 53044, beta-Phellandrene, β-Phellandrene
IUPAC-namn 3-methylidene-6-propan-2-ylcyclohexene
LEDER CC(C)C1CCC(=C)C=C1
Specifikationer
Kemiskt namn eller material beta-Phellandrene
Anteckningar om renhetsgrad HPLC
Kvantitet 1 g
InChI formel InChI=1S/C10H16/c1-8(2)10-6-4-9(3)5-7-10/h4,6,8,10H,3,5,7H2,1-2H3
Formel vikt 136.1252
Procent renhet >95%