Läs mer
Beta-Phenyllongifolol, TRC
Produktkod. 30137067
Change view
Kvantitet:
1 mg
2.5 mg
5 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
30137067 2.5 mg 2.5mg
30135411 1 mg -
30007639 5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30137067 Leverantör Toronto Research Chemicals Leverantörsnummer TRCP3355452.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 946513-36-6
Molekylformel C21H30O
Molekylvikt (g/mol) 298.46
LEDER O[C@H](C1=CC=CC=C1)[C@H]2C3(C)CCCC(C)(C)C4C3CC[C@@H]24
Specifikationer
Kemiskt namn eller material beta-Phenyllongifolol
Anteckningar om renhetsgrad HPLC
Kvantitet 2.5 mg
InChI formel InChI=1S/C21H30O/c1-20(2)12-7-13-21(3)16-11-10-15(17(16)20)18(21)19(22)14-8-5-4-6-9-14/h4-6,8-9,15-19,22H,7,10-13H2,1-3H3/t15-,16?,17?,18+,19-,21?/m1/s1
Formel vikt 298.2297
Procent renhet >95%