Läs mer
(-)-Beta-Sesquiphellandrene, TRC
Produktkod. 17992065
Change view
Kvantitet:
1 mg
2.5 mg
5 mg
Förpackningsstorlek:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
17992065 2.5 mg 2.5mg
17912075 5 mg -
17944735 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 17992065 Leverantör Toronto Research Chemicals Leverantörsnummer TRCS2806552.5MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 20307-83-9
Molekylformel C15 H24
Molekylvikt (g/mol) 204.35
Synonym Cyclohexene, 3-[(1S)-1,5-dimethyl-4-hexen-1-yl]-6-methylene-, (3R)-, 2-Heptene, 2-methyl-6-(4-methylene-2-cyclohexen-1-yl)-, (1'R,6S)-(-)- (8CI), Cyclohexene, 3-(1,5-dimethyl-4-hexenyl)-6-methylene-, [S-(R*,S*)]-, Cyclohexene, 3-[(1S)-1,5-dimethyl-4-hexenyl]-6-methylene-, (3R)- (9CI), β-Sesquiphellandrene (7CI), (3R)-3-[(1S)-1,5-Dimethyl-4-hexen-1-yl]-6-methylenecyclohexene, (-)-(6R,7S)-Sesquiphellandrene, (-)-β-Sesquiphellandrene
IUPAC-namn (3R)-3-[(2S)-6-methylhept-5-en-2-yl]-6-methylidenecyclohexene
LEDER C[C@@H](CCC=C(C)C)[C@H]1CCC(=C)C=C1
Specifikationer
Kemiskt namn eller material beta-Sesquiphellandrene
Kvantitet 2.5 mg
InChI formel InChI=1S/C15H24/c1-12(2)6-5-7-14(4)15-10-8-13(3)9-11-15/h6,8,10,14-15H,3,5,7,9,11H2,1-2,4H3/t14-,15+/m0/s1
Formel vikt 204.1878