Läs mer
Betamethasone 11,21-Diacetate, TRC
Produktkod. 30133922
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30133922 1 g
18053740 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30133922 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB3271001G
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 330157-05-6
Molekylformel C26 H33 F O7
Molekylvikt (g/mol) 476.53
Synonym Betamethasone 11,21-Diacetate, Betamethasone Acetate Imp. C (EP), Pregna-1,4-diene-3,20-dione, 11,21-bis(acetyloxy)-9-fluoro-17-hydroxy-16-methyl-, (11β,16β)-, (11β,16β)-11,21-Bis(acetyloxy)-9-fluoro-17-hydroxy-16-methylpregna-1,4-diene-3,20-dione
IUPAC-namn [2-[(8S,9R,10S,11S,13S,14S,16S,17R)-11-acetyloxy-9-fluoro-17-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] acetate
LEDER C[C@H]1C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@H](C[C@]2(C)[C@@]1(O)C(=O)COC(=O)C)OC(=O)C
Specifikationer
Kemiskt namn eller material Betamethasone 11,21-Diacetate
Kvantitet 1 g
InChI formel InChI=1S/C26H33FO7/c1-14-10-20-19-7-6-17-11-18(30)8-9-23(17,4)25(19,27)22(34-16(3)29)12-24(20,5)26(14,32)21(31)13-33-15(2)28/h8-9,11,14,19-20,22,32H,6-7,10,12-13H2,1-5H3/t14-,19-,20-,22-,23-,24-,25-,26-/m0/s1
Formel vikt 476.221