Läs mer
Betamethasone 17-Butyrate, TRC
Produktkod. 30010691
Change view
Kvantitet:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30010691 100 mg
30006845 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30010691 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB327090100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5534-14-5
Molekylformel C26H35FO6
Molekylvikt (g/mol) 462.55
Synonym 9-Fluoro-11β,17,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione 17-Butyrate;
IUPAC-namn [(8S,9S,10S,11S,13S,16S)-9-fluoro-11-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] butanoate
LEDER CCCC(=O)OC1([C@@H](C)CC2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@]3(F)[C@@H](O)C[C@]12C)C(=O)CO
Specifikationer
Kemiskt namn eller material Betamethasone 17-Butyrate
Analyt- eller komponentnamn Betamethasone 17-Butyrate
Frakt skick Room Temperature
InChI formel InChI=1S/C26H35FO6/c1-5-6-22(32)33-26(21(31)14-28)15(2)11-19-18-8-7-16-12-17(29)9-10-23(16,3)25(18,27)20(30)13-24(19,26)4/h9-10,12,15,18-20,28,30H,5-8,11,13-14H2,1-4H3/t15-,18-,19?,20-,23-,24-,25+,26?/m0/s1
Lösningstyp Neat
Industry Type Forensics
Kvantitet 100 mg
Formel vikt 462.2418