Läs mer
Betamethasone 17-Propionate, TRC
Produktkod. 18061230
Change view
Kvantitet:
2.5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
18061230 25 mg
18050050 2.5 mg
18073520 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 18061230 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB32701525MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5534-13-4
Molekylformel C25 H33 F O6
Molekylvikt (g/mol) 448.52
Synonym 9-Fluoro-11β,21-dihydroxy-16β-methyl-3,20-dioxopregna-1,4-dien-17-yl Propanoate, Betamethasone 17-Propionate, (11β,16β)-9-Fluoro-11,21-dihydroxy-16-methyl-17-(1-oxopropoxy)pregna-1,4-diene-3,20-dione, 9-Fluoro-11β,17,21-trihydroxy-16β-methylpregna-1,4-diene-3,20-dione 17-propionate, Betamethasone 17-propionate, Betamethasone 17α-propionate
IUPAC-namn [(8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11-hydroxy-17-(2-hydroxyacetyl)-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] propanoate
LEDER CCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)CO
Specifikationer
Kemiskt namn eller material Betamethasone 17-Propionate
Anteckningar om renhetsgrad HPLC
Kvantitet 25 mg
InChI formel InChI=1S/C25H33FO6/c1-5-21(31)32-25(20(30)13-27)14(2)10-18-17-7-6-15-11-16(28)8-9-22(15,3)24(17,26)19(29)12-23(18,25)4/h8-9,11,14,17-19,27,29H,5-7,10,12-13H2,1-4H3/t14-,17-,18-,19-,22-,23-,24-,25-/m0/s1
Formel vikt 448.2261
Procent renhet >95%