Läs mer
Betamethasone 21-Acetate 17-Propionate, TRC
Produktkod. 30011409
Change view
Kvantitet:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30011409 1 mg
30009586 10 mg
30010631 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30011409 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB3270401MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 5514-81-8
Molekylformel C27 H35 F O7
Molekylvikt (g/mol) 490.561
Synonym Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17,21-trihydroxy-16β-methyl-, 21-acetate 17-propionate (7CI,8CI), (11β,16β)-21-(Acetyloxy)-9-fluoro-11-hydroxy-16-methyl-17-(1-oxopropoxy)pregna-1,4-diene-3,20-dione, Betamethasone 21-acetate, 17-propionate, 21-(Acetyloxy)-9-fluoro-11β-hydroxy-16β-methyl-3,20-dioxopregna-1,4-dien-17-yl Propanoate, Imp. D (EP)
IUPAC-namn [(8S,9R,10S,11S,13S,14S,16S,17R)-17-(2-acetyloxyacetyl)-9-fluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl] propanoate
LEDER CCC(=O)O[C@@]1([C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C)C(=O)COC(=O)C
Specifikationer
Kemiskt namn eller material Betamethasone 21-Acetate 17-Propionate
Anteckningar om renhetsgrad HPLC
Kvantitet 1 mg
InChI formel InChI=1S/C27H35FO7/c1-6-23(33)35-27(22(32)14-34-16(3)29)15(2)11-20-19-8-7-17-12-18(30)9-10-24(17,4)26(19,28)21(31)13-25(20,27)5/h9-10,12,15,19-21,31H,6-8,11,13-14H2,1-5H3/t15-,19-,20-,21-,24-,25-,26-,27-/m0/s1
Formel vikt 490.2367
Procent renhet >95%