missing translation for 'onlineSavingsMsg'
Läs mer

Betamethasone 21-O-Ethyl Carbonate, TRC

Produktkod. 30136658
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30136658 250 mg
30138155 100 mg
30004007 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 30136658 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB327120250MG

Please to purchase this item. Need a web account? Register with us today!

Denna artikel kan inte returneras. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 52619-05-3
Molekylformel C25 H33 F O7
Molekylvikt (g/mol) 464.52
Synonym Pregna-1,4-diene-3,20-dione, 21-[(ethoxycarbonyl)oxy]-9-fluoro-11,17-dihydroxy-16-methyl-, (11beta,16beta)-, (11beta,16beta)-21-[(Ethoxycarbonyl)oxy]-9-fluoro-11,17-dihydroxy-16-methylpregna-1,4-diene-3,20-dione, Betamethasone EP Impurity D
IUPAC-namn ethyl [2-[(8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] carbonate
LEDER CCOC(=O)OCC(=O)[C@@]1(O)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C

Specifikationer

Kemiskt namn eller material Betamethasone 21-(ethyl carbonate)
Kvantitet 250 mg
InChI formel InChI=1S/C25H33FO7/c1-5-32-21(30)33-13-20(29)25(31)14(2)10-18-17-7-6-15-11-16(27)8-9-22(15,3)24(17,26)19(28)12-23(18,25)4/h8-9,11,14,17-19,28,31H,5-7,10,12-13H2,1-4H3/t14-,17-,18-,19-,22-,23-,24-,25-/m0/s1
Formel vikt 464.221
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.