Läs mer
Betamethasone 21-Valerate, TRC
Produktkod. 30138749
Change view
Kvantitet:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30138749 250 mg
30041268 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 30138749 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB327080250MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 2240-28-0
Molekylformel C27 H37 F O6
Molekylvikt (g/mol) 476.58
Synonym (11β,16β)-9-Fluoro-11,17-dihydroxy-16-methyl-21-[(1-oxopentyl)oxy]-pregna-1,4-diene-3,20-dione, Pregna-1,4-diene-3,20-dione, 9-fluoro-11β,17,21-trihydroxy-16β-methyl-, 21-valerate (7CI,8CI), Betamethasone 21-valerate
IUPAC-namn [2-[(8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-11,17-dihydroxy-10,13,16-trimethyl-3-oxo-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] pentanoate
LEDER CCCCC(=O)OCC(=O)[C@@]1(O)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@@H](O)C[C@]12C
Specifikationer
Kemiskt namn eller material Betamethasone 21-Valerate
Analyt- eller komponentnamn Betamethasone 21-Valerate
Anteckningar om renhetsgrad HPLC
Frakt skick Room Temperature
InChI formel InChI=1S/C27H37FO6/c1-5-6-7-23(32)34-15-22(31)27(33)16(2)12-20-19-9-8-17-13-18(29)10-11-24(17,3)26(19,28)21(30)14-25(20,27)4/h10-11,13,16,19-21,30,33H,5-9,12,14-15H2,1-4H3/t16-,19-,20-,21-,24-,25-,26-,27-/m0/s1
Lösningstyp Neat
Industry Type Pharmaceutical
Kvantitet 250 mg
Formel vikt 476.2574
Procent renhet >95%