Läs mer
Betamethasone 9,11-Epoxide 17,21-Dipropionate, TRC
Produktkod. 30004669
Change view
Kvantitet:
10 mg
50 mg
100 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30004669 10 mg
30008361 100 mg
30006487 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30004669 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB32706010MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 66917-44-0
Molekylformel C28 H36 O7
Molekylvikt (g/mol) 484.58
Synonym 1-(4-Hydroxyphenyl)ethanone Oxime, Paracetamol Imp. G (EP), 4-Hydroxyacetophenone Oxime
IUPAC-namn 4-[(Z)-N-hydroxy-C-methylcarbonimidoyl]phenol
LEDER CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]35O[C@H]5C[C@]12C
Specifikationer
Kemiskt namn eller material Betamethasone 9,11-Epoxide 17,21-Dipropionate
Anteckningar om renhetsgrad HPLC
Kvantitet 10 mg
InChI formel InChI=1S/C28H36O7/c1-6-23(31)33-15-21(30)27(35-24(32)7-2)16(3)12-20-19-9-8-17-13-18(29)10-11-25(17,4)28(19)22(34-28)14-26(20,27)5/h10-11,13,16,19-20,22H,6-9,12,14-15H2,1-5H3/t16-,19-,20-,22-,25-,26-,27-,28+/m0/s1
Formel vikt 484.2461
Procent renhet >95%