Läs mer
Betamethasone dipropionate, MedChemExpress
Produktkod. 16451359
Encompass_Preferred
Change view
Kvantitet:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16451359 1 g
16439855 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Produktkod. 16451359 Leverantör Medchem Express Leverantörsnummer HY13571
MedChemExpress Betamethasone dipropionate is a glucocorticoid steroid with anti-inflammatory and immunosuppressive abilities.
Betamethasone dipropionate is a glucocorticoid steroid with anti-inflammatory and immunosuppressive abilities.
Specifikationer
Kemiskt namn eller material Betamethasone dipropionate
CAS 5593-20-4
Färg White
Anteckningar om renhetsgrad Research
Molekylformel C28H37FO7
Kvantitet 1 g
Synonym Betamethasone 17,21-dipropionate
Löslighetsinformation DMSO : 110 mg/mL (218.00 mM; Need ultrasonic)
LEDER C[C@@]12[C@](C[C@H](C)[C@]2(OC(CC)=O)C(COC(CC)=O)=O)([H])[C@]3([H])CCC4=CC(C=C[C@]4(C)[C@@]3(F)[C@@H](O)C1)=O
Molekylvikt (g/mol) 504.59
Formel vikt 504.59
Procent renhet 98.31%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) COVID-19-immunoregulation