Läs mer
Betamethasone Tripropionate, TRC
Produktkod. 30009220
Change view
Kvantitet:
10 mg
25 mg
50 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
30009220 25 mg
30010351 50 mg
30011318 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Produktkod. 30009220 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB32705025MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiska identifierare
CAS 1186048-33-8
Molekylformel C31 H41 F O8
Molekylvikt (g/mol) 560.65
Synonym (11beta,16beta)-9-Fluoro-16-methyl-11,17,21-tris(1-oxopropoxy)pregna-1,4-diene-3,20-dione, Betamethasone 11,17,21-tripropionate, 9-Fluoro-16beta-methyl-3,20-dioxopregna-1,4-diene-11beta,17,21-triyl Tripropanoate, Betamethasone Tripropionate, Betamethasone Dipropionate Imp. G (Pharmeuropa)
IUPAC-namn [2-[(8S,9R,10S,11S,13S,14S,16S,17R)-9-fluoro-10,13,16-trimethyl-3-oxo-11,17-di(propanoyloxy)-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] propanoate
LEDER CCC(=O)OCC(=O)[C@@]1(OC(=O)CC)[C@@H](C)C[C@H]2[C@@H]3CCC4=CC(=O)C=C[C@]4(C)[C@@]3(F)[C@H](C[C@]12C)OC(=O)CC
Specifikationer
Kemiskt namn eller material Betamethasone Tripropionate
Kvantitet 25 mg
InChI formel InChI=1S/C31H41FO8/c1-7-25(35)38-17-23(34)31(40-27(37)9-3)18(4)14-22-21-11-10-19-15-20(33)12-13-28(19,5)30(21,32)24(16-29(22,31)6)39-26(36)8-2/h12-13,15,18,21-22,24H,7-11,14,16-17H2,1-6H3/t18-,21-,22-,24-,28-,29-,30-,31-/m0/s1
Formel vikt 560.2785