Läs mer
Betulinic acid, MedChemExpress
Beskrivning
Betulinic acid is a natural pentacyclic triterpenoid, acts as a eukaryotic topoisomerase I inhibitor, with an IC50 of 5 μM, and possesses anti-HIV, anti-malarial, anti-inflammatory and anti-tumor properties.
Specifikationer
Specifikationer
| Kemiskt namn eller material | Betulinic acid |
| CAS | 472-15-1 |
| Färg | Off-White |
| Anteckningar om renhetsgrad | Research |
| Molekylformel | C30H48O3 |
| Kvantitet | 100 mg |
| Synonym | Lupatic acid Betulic acid |
| Löslighetsinformation | DMSO : 35.71 mg/mL (78.19 mM; Need ultrasonic) |
| LEDER | [H][C@@]12CC[C@@]3(C)[C@]4(C)CC[C@]5(C(O)=O)[C@@]([C@H](C(C)=C)CC5)([H])[C@@](CC[C@@]([H])3[C@@]1(C)CC[C@H](O)C2(C)C)4[H] |
| Molekylvikt (g/mol) | 456.7 |
| Visa mer |