Learn More
Bexarotene, TRC
Product Code. 30345269
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Product Code. Quantity Unit Size
30345269 50 mg 50mg
1 options
Product Code. 30345269 Supplier Toronto Research Chemicals Supplier No. TRCB33675050MG
 Restricted Item
Beräknad leverans: 10-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Bexarotene
CAS 153559-49-0
Molecular Formula C24 H28 O2
Quantity 50 mg
InChI Formula InChI=1S/C24H28O2/c1-15-13-20-21(24(5,6)12-11-23(20,3)4)14-19(15)16(2)17-7-9-18(10-8-17)22(25)26/h7-10,13-14H,2,11-12H2,1,3-6H3,(H,25,26)
SMILES Cc1cc2c(cc1C(=C)c3ccc(cc3)C(=O)O)C(C)(C)CCC2(C)C
IUPAC Name 4-[1-(3,5,5,8,8-pentamethyl-6,7-dihydronaphthalen-2-yl)ethenyl]benzoic acid
Molecular Weight (g/mol) 348.48
Formula Weight 348.2089