missing translation for 'onlineSavingsMsg'
Läs mer

BG45, TRC

Produktkod. 30346574
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Kvantitet
30346574 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30346574 Supplier Toronto Research Chemicals Supplier No. TRCB3665005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Kemiskt namn eller material N-(2-Aminophenyl)2-pyrazinecarboxamide
CAS 926259-99-6
Molekylformel C11H10N4O
Kvantitet 5 mg
Synonym BG45, N-(2-Aminophenyl)2-pyrazinecarboxamide
InChI formel InChI=1S/C11H10N4O/c12-8-3-1-2-4-9(8)15-11(16)10-7-13-5-6-14-10/h1-7H,12H2,(H,15,16)
LEDER Nc1ccccc1NC(=O)c2cnccn2
IUPAC-namn N-(2-aminophenyl)pyrazine-2-carboxamide
Molekylvikt (g/mol) 214.22
Formel vikt 214.0855
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.