missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BGG Stock Solution, acetylated

The material is tested for level of background fluorescence.

Brand:  Invitrogen™ P2045

Additional Details : Weight : 3.36000kg

Product Code. 11589216

  • 2035.00 SEK / 10mL
Estimated Shipment: 30-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


For Research Use Only. All usage must comply with product instructions.


Description

Description

Invitrogen's Acetylated BGG solution is a 0.5% solution in water.

Specifications

Specifications

10 mL
BGG stock solution is shipped on wet ice and should be stored at 4°C.
Wet Ice
BGG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.