missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ BGLAP/CEN1q FISH Probe
Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.
19880.00 SEK - 32740.00 SEK
Specifications
Content And Storage | Store at 4°C in the dark |
---|---|
Format | Tube |
For Use With (Application) | Fluorescent In Situ Hybridization (Cell) |
Gene | bone gamma-carboxyglutamate (gla) protein |
Gene Alias | BGP,OC,PMF1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16053345
|
Abnova™
FG0214.100UL |
100 μL |
19880.00 SEK
100µL |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16063345
|
Abnova™
FG0214.200UL |
200 μL |
32740.00 SEK
200µL |
Estimated Shipment: 24-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Specifications
Store at 4°C in the dark | |
Fluorescent In Situ Hybridization (Cell) | |
BGP,OC,PMF1 | |
632 | |
Liquid |
Tube | |
bone gamma-carboxyglutamate (gla) protein | |
BGLAP | |
DNA/RNA |