Läs mer
BH3I-1, MedChemExpress
Produktkod. 16489546
Encompass_Preferred
Change view
Kvantitet:
2 mg
5 mg
10 mg
25 mg
50 mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet
16489546 50 mg
16482978 10 mg
16472566 25 mg
16410016 2 mg
16405727 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Produktkod. 16489546 Leverantör Medchem Express Leverantörsnummer HY100383
MedChemExpress BH3I-1 is a Bcl-2 family antagonist, which inhibits the binding of the Bak BH3 peptide to Bcl-xL with a Ki of 2.4±0.2 μM in FP assay. BH3I-1 has a Kd of 5.3 μM against the p53/MDM2 pair.
BH3I-1 is a Bcl-2 family antagonist, which inhibits the binding of the Bak BH3 peptide to Bcl-xL with a Ki of 2.4±0.2 μM in FP assay. BH3I-1 has a Kd of 5.3 μM against the p53/MDM2 pair.
Specifikationer
Kemiskt namn eller material BH3I-1
CAS 300817-68-9
Färg Yellow
Anteckningar om renhetsgrad Research
Molekylformel C15H14BrNO3S2
Kvantitet 50 mg
Synonym BHI1 BH 3I1
Löslighetsinformation DMSO : ≥ 4.1 mg/mL (10.24 mM)
LEDER O=C(O)C(C(C)C)N(C/1=O)C(SC1=C\C2=CC=C(Br)C=C2)=S
Molekylvikt (g/mol) 400.31
Formel vikt 400.31
Procent renhet 98.0%
Kvalitet Research
Fysisk form Solid
För användning med (applikation) Cancer-programmed cell death