Läs mer
BHC, mixture of isomers (a:b:gamma:delta=1:1:1:1), TRC
Produktkod. 30340449
Change view
Kvantitet:
100 mg
Produktkod. Kvantitet
30340449 100 mg
1 options
Produktkod. 30340449 Leverantör Toronto Research Chemicals Leverantörsnummer TRCB372008100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifikationer
Kemiskt namn eller material HCH (BHC) (technical)
CAS 608-73-1
Molekylformel C6 H6 Cl6
Kvantitet 100 mg
Synonym 1,2,3,4,5,6-Hexachlorocyclohexane, BHC, HCH, NSC 11807, NSC 7909, NSC 8093, Hexachlorocyclohexane
InChI formel InChI=1S/C6H6Cl6/c7-1-2(8)4(10)6(12)5(11)3(1)9/h1-6H
LEDER ClC1C(Cl)C(Cl)C(Cl)C(Cl)C1Cl
IUPAC-namn 1,2,3,4,5,6-hexachlorocyclohexane
Molekylvikt (g/mol) 290.83
Formel vikt 287.8601