missing translation for 'onlineSavingsMsg'
Learn More

BHLHE41 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 12688-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16840544

  • 3313.85 SEK / 150µL
Estimated Shipment: 05-10-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

BHLHE41 may be a transcriptional repressor that represses both basal and activated transcription.
Specifications

Specifications

BHLHE41
Polyclonal
Unconjugated
BHLHE41
BHLHB3, BHLHE41, DEC2, hDEC2, SHARP 1, SHARP1
Rabbit
Antigen Affinity Chromatography
RUO
117095, 79362, 79365
-20°C
Liquid
Western Blot, Immunoprecipitation
0.16 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q99PV5, Q9C0J9
Bhlhe41
BHLHE41 Fusion Protein Ag3381
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers