Läs mer
Tocris Bioscience™ BHPI
ERα antagonist; also activates unfolded protein response; active in vivo
Brand: Tocris Bioscience™ 6116/10
Denna artikel kan inte returneras.
Se returpolicy
Beskrivning
BHPI is an estrogen receptor (ER) α inhibitor. Inhibits proliferation of drug-resistant ERα-positive breast and ovarian cancer cells. Induces sustained activation of the unfolded protein response. Active in vivo.Specifikationer
| 1,3-Dihydro-3,3-bis(4-hydroxyphenyl)-7-methyl-2H-indol-2-one | |
| 56632-39-4 | |
| >98% | |
| 10 mg |