missing translation for 'onlineSavingsMsg'
Läs mer

Invitrogen™ Bi-1 Monoclonal Antibody (20F430)

Produktkod. 11559792
Change view
Klicka för att se tillgängliga alternativ
Kvantitet:
100 μg
Förpackningsstorlek:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Kvantitet unitSize
11559792 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11559792 Leverantör Invitrogen™ Leverantörsnummer MA141108

Vänligen för att beställa denna produkten. Behöver du ett Webbkonto? Registrera ditt konto idag!

Denna artikel kan inte returneras. Se returpolicy

Mouse Monoclonal Antibody

Epitope mapping indicates that the antibody recognizes an epitope in the N-terminal region of Bi-1.

BI-1 (Bax Inhibitor 1) is in anti-apoptotic protein that has been linked to protection from apoptosis induced endoplasmic reticulum (ER) stress (reviewed in Bailley-Maitre et al, 2007). BI-1 contains several transmembrane domains, localizes to ER membranes, and has cytoprotective functions that are conserved in both animals and plants. BI-1 suppresses apoptosis induced by ectopic expression of the proapoptotic protein Bax as well as other types of stimuli. Cells from BI-1 knockout (KO) mice are hypersensitivite to apoptosis induced by ER stress-causing chemical agents (thapsigargin, tunicamycin, and brefeldin A) or by ischemia-reperfusion (IR) injury. Conversely, overexpression of BI-1 protects against apoptosis induced by ER stress and IR. The mechanism by which BI-1 protects cells from ER-stress induced apoptosis remains to be fully elucidated, itis thought to involve regulation of Ca2+ handling by the ER.
TRUSTED_SUSTAINABILITY

Specifikationer

Antigen Bi-1
Användningsområden Immunohistochemistry (Paraffin)
Klassificering Monoclonal
Klona 20F430
Koncentration 1.0 mg/mL
Konjugera Unconjugated
Formulering PBS with 0.2% gelatin and 0.05% sodium azide
Gen TMBIM6
Genaccessionsnr. P55061, P55062, Q9D2C7
Gene Alias 5031406P05Rik; Ab1-011; Ac1-149; Bax inhibitor 1; Bax inhibitor-1; BAXI1; Bi1; BI-1; Cc1-27; Tegt; testis enhanced gene transcript; testis-enhanced gene transcript protein; Tmbim6; transmembrane BAX inhibitor motif containing 6; transmembrane BAX inhibitor motif-containing protein 6
Gensymboler TMBIM6
Värd art Mouse
Immunogen A synthetic peptide made to the N-terminal portion of the human Bax inhibitor 1 protein (between amino acids 1-40).
Reningsmetod Protein G
Kvantitet 100 μg
Regulatorisk status RUO
Primär eller sekundär Primary
Gen-ID (Entrez) 110213, 24822, 7009
Målarter Human, Mouse, Rat
Innehåll och lagring Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Produkttyp Antibody
Form Liquid
Isotyp IgG2b κ
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.